Assay Details
| Gene Symbol: LOC100350306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263628.1 | XP_008261850.1 | 8 - 9 | 805 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100350306| Gene Symbol: LOC100350306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263628.1 | XP_008261850.1 | 8 - 9 | 805 | 79 | |
Set Membership: |
|