Assay Details
| Gene Symbol: LOC100352755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342603.1 | XP_017198092.1 | 25 - 26 | 4139 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100352755| Gene Symbol: LOC100352755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342603.1 | XP_017198092.1 | 25 - 26 | 4139 | 91 | |
Set Membership: |
|