Assay Details
| Gene Symbol: TEPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711545.3 | XP_002711591.1 | 6 - 7 | 703 | 74 | |
| XM_008257347.2 | XP_008255569.1 | 5 - 6 | 641 | 74 | ||
More Information
Set Membership: |
|

TEPP
Genomic Map