Assay Details
| Gene Symbol: CIBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255731.2 | XP_008253953.1 | 8 - 9 | 916 | 99 | |
| XM_008255732.2 | XP_008253954.1 | 7 - 8 | 758 | 99 | ||
More Information
Set Membership: |
|

CIBAR1
Genomic Map