Assay Details
| Gene Symbol: LOC100348238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262893.2 | XP_008261115.1 | 20 - 21 | 3126 | 73 | |
| XM_008262894.2 | XP_008261116.1 | 20 - 21 | 3127 | 73 | ||
| XM_008262895.2 | XP_008261117.1 | 20 - 21 | 3352 | 73 | ||
| XM_008262899.2 | XP_008261121.1 | 21 - 22 | 3184 | 73 | ||
| XM_008262901.2 | XP_008261123.1 | 21 - 22 | 3157 | 73 | ||
| XM_017345107.1 | XP_017200596.1 | 20 - 21 | 3126 | 73 | ||
| XM_017345108.1 | XP_017200597.1 | 20 - 21 | 3127 | 73 | ||
| XM_017345109.1 | XP_017200598.1 | 19 - 20 | 2934 | 73 | ||
| XM_017345110.1 | XP_017200599.1 | 20 - 21 | 3130 | 73 | ||
| XM_017345111.1 | XP_017200600.1 | 20 - 21 | 3129 | 73 | ||
| XM_017345112.1 | XP_017200601.1 | 20 - 21 | 3129 | 73 | ||
More Information
Set Membership: |
|

LOC100348238
Genomic Map