Assay Details
| Gene Symbol: LOC100348238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262893.2 | XP_008261115.1 | 5 - 6 | 737 | 108 | |
| XM_008262894.2 | XP_008261116.1 | 5 - 6 | 738 | 108 | ||
| XM_008262895.2 | XP_008261117.1 | 5 - 6 | 963 | 108 | ||
| XM_008262899.2 | XP_008261121.1 | 6 - 7 | 795 | 108 | ||
| XM_008262901.2 | XP_008261123.1 | 6 - 7 | 768 | 108 | ||
| XM_017345107.1 | XP_017200596.1 | 5 - 6 | 737 | 108 | ||
| XM_017345108.1 | XP_017200597.1 | 5 - 6 | 738 | 108 | ||
| XM_017345109.1 | XP_017200598.1 | 5 - 6 | 737 | 108 | ||
| XM_017345110.1 | XP_017200599.1 | 5 - 6 | 741 | 108 | ||
| XM_017345111.1 | XP_017200600.1 | 5 - 6 | 740 | 108 | ||
| XM_017345112.1 | XP_017200601.1 | 5 - 6 | 740 | 108 | ||
More Information
Set Membership: |
|

LOC100348238
Genomic Map