Assay Details
| Gene Symbol: LOC100346422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339590.1 | XP_017195079.1 | 3467 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100346422| Gene Symbol: LOC100346422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339590.1 | XP_017195079.1 | 3467 | 98 | ||
Set Membership: |
|