Assay Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273184.2 | XP_008271406.1 | 13 - 14 | 1557 | 90 | |
| XM_008273186.2 | XP_008271408.1 | 13 - 14 | 1482 | 90 | ||
| XM_017350001.1 | XP_017205490.1 | 13 - 14 | 1504 | 90 | ||
More Information
Set Membership: |
|

ZNF280C
Genomic Map