Assay Details
| Gene Symbol: ATP5MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719309.3 | XP_002719355.1 | 2 - 3 | 128 | 57 | |
| XM_008271336.2 | XP_008269558.1 | 2 - 3 | 106 | 57 | ||
More Information
Set Membership: |
|

ATP5MC1
Genomic Map