Assay Details
| Gene Symbol: TOMM20L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718315.3 | XP_002718361.1 | 2 - 3 | 646 | 120 | |
| XM_017348338.1 | XP_017203827.1 | 2 - 3 | 653 | 120 | ||
More Information
Set Membership: |
|

TOMM20L
Genomic Map