Assay Details
| Gene Symbol: TOMM20L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718315.3 | XP_002718361.1 | 3 - 4 | 733 | 71 | |
| XM_017348337.1 | XP_017203826.1 | 2 - 3 | 858 | 71 | ||
| XM_017348338.1 | XP_017203827.1 | 3 - 4 | 740 | 71 | ||
| XM_017348339.1 | XP_017203828.1 | 2 - 3 | 853 | 71 | ||
| XM_017348340.1 | XP_017203829.1 | 2 - 3 | 879 | 71 | ||
More Information
Set Membership: |
|

TOMM20L
Genomic Map