Assay Details
| Gene Symbol: PDE12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713262.3 | XP_002713308.1 | 1 - 2 | 1990 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDE12| Gene Symbol: PDE12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713262.3 | XP_002713308.1 | 1 - 2 | 1990 | 115 | |
Set Membership: |
|