Assay Details
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710010.3 | XP_002710056.1 | 1 - 2 | 153 | 66 | |
XM_008253270.2 | XP_008251492.1 | 1 - 2 | 153 | 66 | ||
XM_017339478.1 | XP_017194967.1 | 1 - 2 | 153 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
