Assay Details
| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714566.3 | XP_002714612.1 | 6 - 7 | 819 | 67 | |
| XM_008263191.2 | XP_008261413.1 | 5 - 6 | 608 | 67 | ||
More Information
Set Membership: |
|

SLC35A1
Genomic Map