Assay Details
| Gene Symbol: LRRC74A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349426.1 | XP_017204915.1 | 7 - 8 | 737 | 105 | |
| XM_017349427.1 | XP_017204916.1 | 7 - 8 | 737 | 105 | ||
| XM_017349428.1 | XP_017204917.1 | 7 - 8 | 737 | 105 | ||
More Information
Set Membership: |
|

LRRC74A
Genomic Map