Assay Details
| Gene Symbol: LOC100340535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273104.2 | XP_008271326.1 | 8 - 9 | 972 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100340535| Gene Symbol: LOC100340535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273104.2 | XP_008271326.1 | 8 - 9 | 972 | 88 | |
Set Membership: |
|