Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259394.2 | XP_008257616.1 | 2 - 3 | 186 | 72 | |
| XM_008259395.2 | XP_008257617.1 | 2 - 3 | 186 | 72 | ||
| XM_017343269.1 | XP_017198758.1 | 2 - 3 | 402 | 72 | ||
More Information
Set Membership: |
|

YAF2
Genomic Map