Assay Details
| Gene Symbol: KANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722017.3 | XP_002722063.1 | 2511 | 61 | ||
| XM_008249138.2 | XP_008247360.1 | 2429 | 61 | |||
| XM_008249139.1 | XP_008247361.1 | 2494 | 61 | |||
| XM_008249140.1 | XP_008247362.1 | 2470 | 61 | |||
More Information
Set Membership: |
|

KANK3
Genomic Map