Assay Details
| Gene Symbol: LOC100339421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719363.3 | XP_002719409.1 | 6 - 7 | 1184 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100339421| Gene Symbol: LOC100339421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719363.3 | XP_002719409.1 | 6 - 7 | 1184 | 88 | |
Set Membership: |
|