Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267433.2 | XP_008265655.2 | 3 - 4 | 345 | 98 | |
XM_017347339.1 | XP_017202828.1 | 4 - 5 | 516 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
