Assay Details
| Gene Symbol: GPR89B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715650.3 | XP_002715696.1 | 9 - 10 | 944 | 74 | |
| XM_008264554.2 | XP_008262776.1 | 9 - 10 | 958 | 74 | ||
More Information
Set Membership: |
|

GPR89B
Genomic Map