Assay Details
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722932.3 | XP_002722978.2 | 602 | 77 | ||
| XM_017338904.1 | XP_017194393.1 | 606 | 77 | |||
| XM_017338905.1 | XP_017194394.1 | 463 | 77 | |||
More Information
Set Membership: |
|

CDK3
Genomic Map