Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711089.3 | XP_002711135.1 | 11 - 12 | 980 | 70 | |
| XM_008256409.2 | XP_008254631.1 | 10 - 11 | 870 | 70 | ||
More Information
Set Membership: |
|

PRKAG1
Genomic Map