Assay Details
| Gene Symbol: LOC100352221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257007.2 | XP_008255229.1 | 12 - 13 | 1247 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100352221| Gene Symbol: LOC100352221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257007.2 | XP_008255229.1 | 12 - 13 | 1247 | 63 | |
Set Membership: |
|