Assay Details
| Gene Symbol: BLVRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261676.2 | XP_008259898.1 | 8 - 9 | 865 | 58 | |
| XM_008261677.2 | XP_008259899.1 | 7 - 8 | 832 | 58 | ||
| XM_017344453.1 | XP_017199942.1 | 6 - 7 | 779 | 58 | ||
More Information
Set Membership: |
|

BLVRA
Genomic Map