Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266506.2 | XP_008264728.1 | 12 - 13 | 2111 | 103 | |
| XM_008266521.2 | XP_008264743.1 | 14 - 15 | 2282 | 103 | ||
| XM_017346923.1 | XP_017202412.1 | 11 - 12 | 2016 | 103 | ||
More Information
Set Membership: |
|

NAALADL2
Genomic Map