Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266506.2 | XP_008264728.1 | 3 - 4 | 942 | 67 | |
| XM_008266521.2 | XP_008264743.1 | 5 - 6 | 1113 | 67 | ||
| XM_017346923.1 | XP_017202412.1 | 3 - 4 | 943 | 67 | ||
More Information
Set Membership: |
|

NAALADL2
Genomic Map