Assay Details
| Gene Symbol: PPIH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265449.2 | XP_008263671.1 | 5 - 6 | 293 | 68 | |
| XM_008265450.2 | XP_008263672.1 | 5 - 6 | 293 | 68 | ||
| XM_008265451.2 | XP_008263673.1 | 5 - 6 | 293 | 68 | ||
| XM_008265452.2 | XP_008263674.1 | 5 - 6 | 293 | 68 | ||
| XM_008265453.2 | XP_008263675.1 | 5 - 6 | 293 | 68 | ||
More Information
Set Membership: |
|

PPIH
Genomic Map