Assay Details
| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723988.2 | XP_002724034.2 | 431 | 69 | ||
| XM_017340110.1 | XP_017195599.1 | 431 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002723988.2, XM_017340110.1 transcripts.
Set Membership: |
|

SIGLECL1
Genomic Map