Assay Details
| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723988.2 | XP_002724034.2 | 903 | 96 | ||
| XM_017340110.1 | XP_017195599.1 | 903 | 96 | |||
| XM_017340111.1 | XP_017195600.1 | 715 | 96 | |||
More Information
Set Membership: |
|

SIGLECL1
Genomic Map