Assay Details
| Gene Symbol: ACP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709090.3 | XP_002709136.1 | 8 - 9 | 893 | 55 | |
| XM_017346419.1 | XP_017201908.1 | 8 - 9 | 895 | 55 | ||
More Information
Set Membership: |
|

ACP2
Genomic Map