Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711058.3 | XP_002711104.1 | 2 - 3 | 387 | 69 | |
| XM_008256608.2 | XP_008254830.1 | 2 - 3 | 383 | 69 | ||
| XM_017341924.1 | XP_017197413.1 | 1 - 2 | 501 | 69 | ||
More Information
Set Membership: |
|

ORMDL2
Genomic Map