Assay Details
| Gene Symbol: ERICH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716264.2 | XP_002716310.1 | 4 - 5 | 624 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH6| Gene Symbol: ERICH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716264.2 | XP_002716310.1 | 4 - 5 | 624 | 76 | |
Set Membership: |
|