Assay Details
| Gene Symbol: LOC100344331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265588.1 | XP_008263810.1 | 9 - 10 | 1315 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100344331| Gene Symbol: LOC100344331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265588.1 | XP_008263810.1 | 9 - 10 | 1315 | 74 | |
Set Membership: |
|