Assay Details
| Gene Symbol: LOC103351797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272568.1 | XP_008270790.1 | 1 - 2 | 102 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008272568.1 transcripts.
Set Membership: |
|

LOC103351797
Genomic Map