Assay Details
| Gene Symbol: LOC103351797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272568.1 | XP_008270790.1 | 8 - 9 | 789 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103351797| Gene Symbol: LOC103351797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272568.1 | XP_008270790.1 | 8 - 9 | 789 | 72 | |
Set Membership: |
|