Assay Details
| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717124.3 | XP_002717170.1 | 2 - 3 | 927 | 73 | |
| XM_008267780.2 | XP_008266002.1 | 2 - 3 | 863 | 73 | ||
More Information
Set Membership: |
|

UGT2A1
Genomic Map