Assay Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267308.2 | XP_008265530.1 | 3 - 4 | 872 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBM46| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267308.2 | XP_008265530.1 | 3 - 4 | 872 | 95 | |
Set Membership: |
|