Assay Details
| Gene Symbol: MINDY4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346671.1 | XP_017202160.1 | 3 - 4 | 174 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MINDY4B| Gene Symbol: MINDY4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346671.1 | XP_017202160.1 | 3 - 4 | 174 | 84 | |
Set Membership: |
|