Assay Details
| Gene Symbol: LOC100353262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342512.1 | XP_017198001.1 | 23 - 24 | 4296 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100353262| Gene Symbol: LOC100353262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342512.1 | XP_017198001.1 | 23 - 24 | 4296 | 116 | |
Set Membership: |
|