Assay Details
| Gene Symbol: LOC103345049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337615.1 | XP_017193104.1 | 1553 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103345049| Gene Symbol: LOC103345049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337615.1 | XP_017193104.1 | 1553 | 115 | ||
Set Membership: |
|