Assay Details
| Gene Symbol: LOC103345049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337615.1 | XP_017193104.1 | 125 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017337615.1 transcripts.
Set Membership: |
|

LOC103345049
Genomic Map