Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708805.3 | XP_002708851.1 | 9 - 10 | 1738 | 71 | |
| XM_008266070.2 | XP_008264292.1 | 9 - 10 | 1168 | 71 | ||
| XM_017345326.1 | XP_017200815.1 | 9 - 10 | 1089 | 71 | ||
More Information
Set Membership: |
|

WEE1
Genomic Map