Assay Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716339.3 | XP_002716385.1 | 1 - 2 | 368 | 93 | |
| XM_008266413.2 | XP_008264635.1 | 1 - 2 | 369 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002716339.3, XM_008266413.2 transcripts.
Set Membership: |
|

TRIM59
Genomic Map