Assay Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716339.3 | XP_002716385.1 | 2 - 3 | 444 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002716339.3 transcripts.
Set Membership: |
|

TRIM59
Genomic Map