Assay Details
| Gene Symbol: FBXL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270409.2 | XP_008268631.1 | 4 - 5 | 753 | 63 | |
| XM_017348521.1 | XP_017204010.1 | 3 - 4 | 2019 | 63 | ||
More Information
Set Membership: |
|

FBXL15
Genomic Map