Assay Details
| Gene Symbol: LOC103346006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251392.2 | XP_008249614.2 | 4632 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103346006| Gene Symbol: LOC103346006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251392.2 | XP_008249614.2 | 4632 | 100 | ||
Set Membership: |
|