Assay Details
| Gene Symbol: LOC103346006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251392.2 | XP_008249614.2 | 3292 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008251392.2 transcripts.
Set Membership: |
|

LOC103346006
Genomic Map