Assay Details
| Gene Symbol: PWWP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272838.2 | XP_008271060.1 | 2 - 3 | 203 | 67 | |
| XM_008272839.2 | XP_008271061.1 | 2 - 3 | 215 | 67 | ||
| XM_008272840.2 | XP_008271062.1 | 2 - 3 | 228 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008272838.2, XM_008272839.2, XM_008272840.2 transcripts.
Set Membership: |
|

PWWP3B
Genomic Map