Assay Details
| Gene Symbol: SMIM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249628.2 | XP_008247850.1 | 476 | 80 | ||
| XM_008249632.2 | XP_008247854.1 | 267 | 80 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM22| Gene Symbol: SMIM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249628.2 | XP_008247850.1 | 476 | 80 | ||
| XM_008249632.2 | XP_008247854.1 | 267 | 80 | |||
Set Membership: |
|